Learn More
Medchemexpress LLC Losartan-d4 (carboxylic acid) | 1246820-62-1 | 98.1% | 1 MG
SDP
Catalog No. 5000341692
Encompass_Preferred
Losartan-d4 carboxylic acid (E-3174-d4) is a deuterium-labeled Losartan Carboxylic Acid. It is an active carboxylic acid metabolite of Losartan and an angiotensin II receptor type 1 (AT1) antagonist. It blocks angiotensin II-induced responses in vascular smooth muscle cells (VSMC) and elevates plasma renin activities while reducing mean arterial pressure.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration can affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-003-41692 Supplier Medchemexpress LLC Supplier No. HY12765S1MG
Losartan-d4 carboxylic acid (E-3174-d4) is a deuterium-labeled Losartan Carboxylic Acid. It is an active carboxylic acid metabolite of Losartan and an angiotensin II receptor type 1 (AT1) antagonist. It blocks angiotensin II-induced responses in vascular smooth muscle cells (VSMC) and elevates plasma renin activities while reducing mean arterial pressure.
  • Used as a tracer.
  • Used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration can affect the pharmacokinetic and metabolic profiles of drugs.