Learn More
Apexbio Technology LLC Losartan Potassium (DuP 753) 124750-99-8 10mM (in 1mL DMSO)
SDP
Catalog No. 5000063213
Encompass_Preferred
Losartan Potassium (DuP 753 CAS 124750-99-8) is a small molecule antagonist selective toward angiotensin AT receptors By competitively blocking AT receptors it inhibits angiotensin II-mediated vasoconstriction demonstrated by antagonism of contractile responses in isolated rabbit aortic and jugular vein tissues (pA 8 27) As an orally bioavailable agent losartan potassium is frequently utilized in cardiovascular research notably in studies investigating hypertension and associated mechanisms of angiotensin II signaling
Catalog No. 50-000-63213 Supplier Apexbio Technology LLC Supplier No. A14255.1
Losartan Potassium (DuP 753 CAS 124750-99-8) is a small molecule antagonist selective toward angiotensin AT receptors By competitively blocking AT receptors it inhibits angiotensin II-mediated vasoconstriction demonstrated by antagonism of contractile responses in isolated rabbit aortic and jugular vein tissues (pA 8 27) As an orally bioavailable agent losartan potassium is frequently utilized in cardiovascular research notably in studies investigating hypertension and associated mechanisms of angiotensin II signaling