Learn More
Cayman Chemical Losartn-d4 500ug
SDP
Catalog No. 502986740
Encompass_Preferred
An internal standard for the quantification of losartan (potassium salt) by GC- or LC-MS
Catalog No. 50-298-6740 Supplier Cayman Chemical Supplier No. 22567500 UG
An internal standard for the quantification of losartan (potassium salt) by GC- or LC-MS