Learn More
Medchemexpress LLC HY-10402 50mg , Losmapimod GSK-AHAB;GW856553X;SB856553 CAS:585543-15-3 Purity:98%
SDP
Catalog No. 501493708
Encompass_Preferred
Medchemexpress, HY-10402 50mg Losmapimod GSK-AHAB;GW856553X;SB856553 CAS:585543-15-3 Formula:C22H26FN3O2 pKi: 8.1 (p38α), 7.6 (p38β) Purity:98% Losmapimod is a selective, potent, and orally active p38 MAPK inhibitor with pK i of 8.1 and 7.6 for p38α and p38β, respectively. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-149-3708 Supplier Medchemexpress LLC Supplier No. HY1040250MG
Medchemexpress, HY-10402 50mg Losmapimod GSK-AHAB;GW856553X;SB856553 CAS:585543-15-3 Formula:C22H26FN3O2 pKi: 8.1 (p38α), 7.6 (p38β) Purity:98% Losmapimod is a selective, potent, and orally active p38 MAPK inhibitor with pK i of 8.1 and 7.6 for p38α and p38β, respectively. Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.