Learn More
Cayman Chemical Loteprednol etabonate, Purity: ≥98%, CAS: 82034-46-6, 10 mg
SDP
Catalog No. 502993844
Encompass_Preferred
A glucocorticoid receptor agonist with an affinity 4.3 times that of dexamethasone; minimizes scarring of rabbit cornea wounds
Catalog No. 50-299-3844 Supplier Cayman Chemical Supplier No. 2330510 MG
A glucocorticoid receptor agonist with an affinity 4.3 times that of dexamethasone; minimizes scarring of rabbit cornea wounds