Learn More
Selleck Chemical LLC Loteprednol etabonate 10mg 82034-46-6
SDP
Supplier:  Selleck Chemical LLC S166910MG
Encompass_Preferred
Loteprednol etabonate is a potent glucocorticoid receptor agonist, used in treatment of inflammation of the eye due to allergies. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-597-5