Learn More
Cayman Chemical LotIglIpronhydrochlorIde 10mg
SDP
Catalog No. 502945046
Encompass_Preferred
A GLP-1R agonist; induces cAMP accumulation in cells expressing human GLP-1R (EC50 = 0.96 nM)
Catalog No. 50-294-5046 Supplier Cayman Chemical Supplier No. 4122710 MG
A GLP-1R agonist; induces cAMP accumulation in cells expressing human GLP-1R (EC50 = 0.96 nM)