Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Lotilaner 25mg | 1369852-71-0 | 596.76 g/mol | C20H14Cl3F6N3O3S | 25 MG
SDP

Catalog No. 5000227266
Encompass_Preferred

Lotilaner is an isoxazoline-class parasiticide used in research to target ectoparasites by antagonizing GABA-gated chloride channels. It is supplied as a high-purity research chemical for in vitro and in vivo studies.

  • Parasiticide acting as a non-competitive antagonist of insect GABA-gated chloride receptors.
  • CAS 1369852-71-0; molecular formula C20H14Cl3F6N3O3S; molecular weight 596.8 g/mol.
  • High purity (99.84%), suitable for analytical and biological research.
  • Soluble in DMSO at ≥100 mg/mL; in vivo formulations reported at ≥2.5 mg/mL in mixed vehicles.
  • Recommended storage: powder at -20°C (long term) or 4°C; solutions at -80°C or -20°C depending on duration.

Catalog No. 50-002-27266 Supplier Medchemexpress LLC Supplier No. HY11656425MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Lotilaner is an isoxazoline-class parasiticide used in research to target ectoparasites by antagonizing GABA-gated chloride channels. It is supplied as a high-purity research chemical for in vitro and in vivo studies.

  • Parasiticide acting as a non-competitive antagonist of insect GABA-gated chloride receptors.
  • CAS 1369852-71-0; molecular formula C20H14Cl3F6N3O3S; molecular weight 596.8 g/mol.
  • High purity (99.84%), suitable for analytical and biological research.
  • Soluble in DMSO at ≥100 mg/mL; in vivo formulations reported at ≥2.5 mg/mL in mixed vehicles.
  • Recommended storage: powder at -20°C (long term) or 4°C; solutions at -80°C or -20°C depending on duration.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.