Learn More
R&R Lotions LOTION CR-ESD W/ALOE 16OZ PMP
SDP
Catalog No. NC9453626
Encompass_Preferred
16 oz. I.C. Antistatic Cleanroom Lotion is Fragrance and Dye Free and is enriched with Aloe Vera & Vitamins AD & E. This lotion is bottled in our ESD Static Dissipative container with an average surface resistivity of 10^9 to 10^10 Ohm/sq. and will dissipate a static charge of +/- 5000V in less than
Catalog No. NC9453626 Supplier R&R Lotions Supplier No. ICL16CRESD
16 oz. I.C. Antistatic Cleanroom Lotion is Fragrance and Dye Free and is enriched with Aloe Vera & Vitamins AD & E. This lotion is bottled in our ESD Static Dissipative container with an average surface resistivity of 10^9 to 10^10 Ohm/sq. and will dissipate a static charge of +/- 5000V in less than