Learn More
ACL Inc LOTION ESD SAFE 24/CS
Catalog No. 501037864
Encompass_Preferred
This static dissipative hand lotion is equipped to prevent and handle static discharge which is especially important in the electronic's industry. Contains no glycerin, silicone, lanolin or mineral oil. Complies with DOD 1686A and DOD HDBK 263.
Catalog No. 50-103-7864 Supplier ACL Inc Supplier No. 7001
This static dissipative hand lotion is equipped to prevent and handle static discharge which is especially important in the electronic's industry. Contains no glycerin, silicone, lanolin or mineral oil. Complies with DOD 1686A and DOD HDBK 263.