Learn More
Vector Laboratories Lotus Tetragonolobus Lectin (LTL) Biotinylated 2 mg
SDP
Catalog No. NC0370187
Encompass
Lotus tetragonolobus lectin (LTL) encompasses a family of closely related glycoproteins with similar specificities toward α-linked L-fucose-containing oligosaccharides. Although many of the binding properties of Lotus lectin are similar to those of Ulex europaeus lectin I (UEL I), the binding affinities and some specificities for oligosaccharides are significantly different between these fucose-specific lectins. This conjugate features a ratio of biotin to lectin protein that provides optimal avidin/streptavidin and lectin binding activity. Supplied essentially free of unconjugated biotins, it is preserved with sodium azide.
Catalog No. NC0370187 Supplier Vector Laboratories Supplier No. B13252
Lotus tetragonolobus lectin (LTL) encompasses a family of closely related glycoproteins with similar specificities toward α-linked L-fucose-containing oligosaccharides. Although many of the binding properties of Lotus lectin are similar to those of Ulex europaeus lectin I (UEL I), the binding affinities and some specificities for oligosaccharides are significantly different between these fucose-specific lectins. This conjugate features a ratio of biotin to lectin protein that provides optimal avidin/streptavidin and lectin binding activity. Supplied essentially free of unconjugated biotins, it is preserved with sodium azide.