Learn More
FOSSIL ION TECHNOLOGY SL LOTUS TIP 20UM ID 363UM OD
Catalog No. NC2224743
Encompass
Fossil Ion Tech LOTUS tips Size: L-20-04 -20um ID, 363um OD, 4cm Long, 15 degrees Sharp
This item is not returnable. View return policy
Catalog No. NC2224743 Supplier FOSSIL ION TECHNOLOGY SL Supplier No. L2004
Fossil Ion Tech LOTUS tips Size: L-20-04 -20um ID, 363um OD, 4cm Long, 15 degrees Sharp