Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Lotusine hydroxide | 3721-76-4 | 99.8% | 331.41 g/mol | C19H25NO4 | 10 MG
SDP

Catalog No. 5000285312
Encompass_Preferred

Lotusine hydroxide is a natural benzyltetrahydroisoquinoline alkaloid isolated from the green seed embryo of Nelumbo nucifera. Supplied as a solid for in vitro pharmacology and electrophysiology research, it is documented to affect myocardial action potentials and is soluble in DMSO with hygroscopic considerations.

  • High purity (99.81%) for reproducible in vitro results.
  • Molecular weight 331.41 g/mol and formula C19H25NO4 for compound reference.
  • Provided as a 10 MG solid suitable for small-scale research.
  • Soluble in DMSO (33.33 mg/mL) with ultrasonic warming recommended for dissolution.
  • COA and SDS available for quality and safety documentation.

Catalog No. 50-002-85312 Supplier Medchemexpress LLC Supplier No. HYN4309A10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Lotusine hydroxide is a natural benzyltetrahydroisoquinoline alkaloid isolated from the green seed embryo of Nelumbo nucifera. Supplied as a solid for in vitro pharmacology and electrophysiology research, it is documented to affect myocardial action potentials and is soluble in DMSO with hygroscopic considerations.

  • High purity (99.81%) for reproducible in vitro results.
  • Molecular weight 331.41 g/mol and formula C19H25NO4 for compound reference.
  • Provided as a 10 MG solid suitable for small-scale research.
  • Soluble in DMSO (33.33 mg/mL) with ultrasonic warming recommended for dissolution.
  • COA and SDS available for quality and safety documentation.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.