Learn More
ATCC Loucy; Acute T Cell Leukemia; Human (Homo sapiens)
Catalog No. 502384161
Encompass
Loucy is a T lymphoblast cell line that was isolated in 1987 from the peripheral blood of a 38-year-old, White female patient with T- cell acute lymphoblastic leukemia . This cell line was deposited by H Ben-Bassatand can be used in immunology and immune system disorder research.
Catalog No. 50-238-4161 Supplier ATCC Supplier No. CRL2629
Loucy is a T lymphoblast cell line that was isolated in 1987 from the peripheral blood of a 38-year-old, White female patient with T- cell acute lymphoblastic leukemia . This cell line was deposited by H Ben-Bassatand can be used in immunology and immune system disorder research.