Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Supertek™ Loud Speaker Kit
GREENER_CHOICESCI_ED

Catalog No. S160135 Shop All Supertek Scientific Products
Change view
Click to view available options
Grade:
6 to 12
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade
S160135 6 to 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S160135 Supplier Supertek™ Supplier No. PH36210
Add to Cart
Add to Cart
This item is not returnable. View return policy

Illustrate how electric signals are converted into sound in loud speakers by using the Supertek™ Loud Speaker Kit.

  • Super fun way to learn!
  • Illustrates how electric signals are converted into sound in loud speakers.
  • This kit can also be used to explain electromagnetism, the interaction of magnetic fields and the mechanical amplification of sound.
  • Components shown in picture are included.
  • Comes with companion guide.

Specifications

Grade 6 to 12
Format Kit
Languages English
Subject Physics - Sound and Light
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.