Learn More
MSC Louisville Ladder Step Stool, Fiberglass, 1 foot, Type IAA, 375 lb
Catalog No. 5000513602
Encompass
This industrial step stool is constructed from lightweight fiberglass, offering durability and stability for various work environments. Designed with a 1-foot standing height and a robust 375 lb load capacity, it provides a secure platform for tasks requiring elevated access. This step stool meets or exceeds safety standards set by ANSI and OSHA.
  • Large molded top provides a spacious working surface.
  • Large aluminum steps ensure secure footing.
  • Double-rivet step construction enhances durability.
  • Heavy-duty gusset bracing adds structural integrity.
Catalog No. 50-005-13602 Supplier MSC Supplier No. 00236455
This industrial step stool is constructed from lightweight fiberglass, offering durability and stability for various work environments. Designed with a 1-foot standing height and a robust 375 lb load capacity, it provides a secure platform for tasks requiring elevated access. This step stool meets or exceeds safety standards set by ANSI and OSHA.
  • Large molded top provides a spacious working surface.
  • Large aluminum steps ensure secure footing.
  • Double-rivet step construction enhances durability.
  • Heavy-duty gusset bracing adds structural integrity.