Learn More
Roboz Surgical Loupes on Sports Eyeglasses, 5X magnification, 420mm working distance
SDP
Catalog No. NC1315898
Encompass
Loupes on Sports Eyeglasses, 5X magnification, 420mm working distance. 60mm Field of view, 80mm depth of field.
This item is not returnable. View return policy
Catalog No. NC1315898 Supplier Roboz Surgical Supplier No. ML643
Loupes on Sports Eyeglasses, 5X magnification, 420mm working distance. 60mm Field of view, 80mm depth of field.