Learn More
Medchemexpress LLC Loureirin A | 119425-89-7 | 286.32 | 1 MG
SDP
Catalog No. 5000415608
Encompass_Preferred
Loureirin A is a flavonoid extracted from Dragon's Blood. It functions as an inhibitor of Akt phosphorylation and exhibits antiplatelet activity.
  • Inhibits Akt phosphorylation.
  • Possesses antiplatelet activity.
Catalog No. 50-004-15608 Supplier Medchemexpress LLC Supplier No. HYN15051MG
Loureirin A is a flavonoid extracted from Dragon's Blood. It functions as an inhibitor of Akt phosphorylation and exhibits antiplatelet activity.
  • Inhibits Akt phosphorylation.
  • Possesses antiplatelet activity.