Learn More
Medchemexpress LLC Loureirin A | 119425-89-7 | 286.32 | 1 ML
SDP
Catalog No. 5000415280
Encompass_Preferred
Loureirin A is a flavonoid extracted from Dragon's Blood. It is known to inhibit Akt phosphorylation and possesses antiplatelet activity. It is for research use only and not sold to patients.
  • Inhibits Akt phosphorylation.
  • Has antiplatelet activity.
  • Extracted from Dragon's Blood.
  • For research use only.
Catalog No. 50-004-15280 Supplier Medchemexpress LLC Supplier No. HYN150510MM/1ML
Loureirin A is a flavonoid extracted from Dragon's Blood. It is known to inhibit Akt phosphorylation and possesses antiplatelet activity. It is for research use only and not sold to patients.
  • Inhibits Akt phosphorylation.
  • Has antiplatelet activity.
  • Extracted from Dragon's Blood.
  • For research use only.