Learn More
Selleck Chemical LLC Loureirin A S3241-1mg
SDP
Catalog No. 503110859
Encompass_Preferred
Loureirin A is a flavonoid extracted from the red resin of the herbs of Dracaena cochinchinensis which is known as Dragon s Blood Loureirin A inhibits platelet activation by an impairment of PI3K/Akt signaling Loureirin A inhibits Akt phosphorylation
Catalog No. 50-311-0859 Supplier Selleck Chemical LLC Supplier No. S32411MG
Loureirin A is a flavonoid extracted from the red resin of the herbs of Dracaena cochinchinensis which is known as Dragon s Blood Loureirin A inhibits platelet activation by an impairment of PI3K/Akt signaling Loureirin A inhibits Akt phosphorylation