Learn More
TARGETMOL CHEMICALS INC LOUREIRIN A 5MG
SDP
Catalog No. 502579716
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Loureirin A has an inhibitory effect on platelet activation perhaps through an impairment of PI3K/Akt signaling.Loureirin A negatively affects agonist-induced platelet aggregation such as collagen collagen-related peptide (CRP) ADP and thrombin. Loureirin A activates Wnt/-catenin pathway and promotes hair follicle stem cells (FSCs)-seeded tissue-engineered skin to repair skin wound.The molecular mechanism of Loureirin A and Wnt signaling pathway mediated anti- hepatic fibrosis and anti- angiogenesis may involve down- regulation the expression of Frizzled- 4inhibiting the synthesis and secretion of (alpha)- SMATGF- (beta)1and the proliferation of HSCs. purity: 99%
Catalog No. 50-257-9716 Supplier TARGETMOL CHEMICALS INC Supplier No. T5S08965MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Loureirin A has an inhibitory effect on platelet activation perhaps through an impairment of PI3K/Akt signaling.Loureirin A negatively affects agonist-induced platelet aggregation such as collagen collagen-related peptide (CRP) ADP and thrombin. Loureirin A activates Wnt/-catenin pathway and promotes hair follicle stem cells (FSCs)-seeded tissue-engineered skin to repair skin wound.The molecular mechanism of Loureirin A and Wnt signaling pathway mediated anti- hepatic fibrosis and anti- angiogenesis may involve down- regulation the expression of Frizzled- 4inhibiting the synthesis and secretion of (alpha)- SMATGF- (beta)1and the proliferation of HSCs. purity: 99%