Learn More
Cayman Chemical Loureirin B, Purity: ≥98%, CAS: 119425-90-0, 10 mg
SDP
Catalog No. 502940528
Encompass_Preferred
A flavonoid with diverse biological activities; inhibits Kv1.3-mediated currents, induces membrane depolarization, and reduces calcium influx in Jurkat T cells; inhibits phytohemagglutinin-induced IL-2 release in Jurkat T cells; reduces type I collagen and fibronectin protein levels in TGF-β1-stimulated fibroblasts as well as contraction of TGF-β1-stimulated fibroblasts in a gel contraction assay at 25 μg/ml; reduces type I collagen and fibronectin protein levels and inhibits phosphorylation of ERK and JNK in isolated human hypertrophic scar tissue; increases glucose absorption and intracellular ATP levels in Ins-1 cells
Catalog No. 50-294-0528 Supplier Cayman Chemical Supplier No. 2745410 MG
A flavonoid with diverse biological activities; inhibits Kv1.3-mediated currents, induces membrane depolarization, and reduces calcium influx in Jurkat T cells; inhibits phytohemagglutinin-induced IL-2 release in Jurkat T cells; reduces type I collagen and fibronectin protein levels in TGF-β1-stimulated fibroblasts as well as contraction of TGF-β1-stimulated fibroblasts in a gel contraction assay at 25 μg/ml; reduces type I collagen and fibronectin protein levels and inhibits phosphorylation of ERK and JNK in isolated human hypertrophic scar tissue; increases glucose absorption and intracellular ATP levels in Ins-1 cells