Learn More
TARGETMOL CHEMICALS INC LOUREIRIN B 5MG
SDP
Catalog No. 502580086
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Loureirin B can downregulate the expression of fibrosis-related molecules by regulating MMPs and TIMPs levels inhibit scar fibroblast proliferation and suppress TGF-(beta)1-induced fibrosis during which TGF-(beta)1/Smad2/3 pathway is likely involved. purity: 99%
Catalog No. 50-258-0086 Supplier TARGETMOL CHEMICALS INC Supplier No. T38765MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Loureirin B can downregulate the expression of fibrosis-related molecules by regulating MMPs and TIMPs levels inhibit scar fibroblast proliferation and suppress TGF-(beta)1-induced fibrosis during which TGF-(beta)1/Smad2/3 pathway is likely involved. purity: 99%