Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Loureirin D | 119425-91-1 | 5 MG
SDP

Catalog No. 5000420042
Encompass_Preferred

Loureirin D is a dihydrochalcone extracted from dragon's blood, intended for research use only.

  • Purity: 99.30%
  • Molecular weight: 288.30
  • Formula: C16H16O5
  • Appearance: Solid, white to pink
  • Soluble in DMSO (≥ 50 mg/mL)
  • Storage conditions: 4°C, protected from light; in solvent: -80°C for 6 months, -20°C for 1 month (protected from light)
  • Structure classification: Phenols, polyphenols
  • Initial source: Plants (other families)

Catalog No. 50-004-20042 Supplier Medchemexpress LLC Supplier No. HYN81895MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Loureirin D is a dihydrochalcone extracted from dragon's blood, intended for research use only.

  • Purity: 99.30%
  • Molecular weight: 288.30
  • Formula: C16H16O5
  • Appearance: Solid, white to pink
  • Soluble in DMSO (≥ 50 mg/mL)
  • Storage conditions: 4°C, protected from light; in solvent: -80°C for 6 months, -20°C for 1 month (protected from light)
  • Structure classification: Phenols, polyphenols
  • Initial source: Plants (other families)

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.