Learn More
Medchemexpress LLC HY-N0504 100mg , Lovastatin CAS:75330-75-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN0504100MG
Encompass_Preferred
HY-N0504 100mg Lovastatin CAS: 75330-75-5 Lovastatin is a cell-permeable HMG-CoA reductase inhibitor used to lower cholesterol. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0311