Learn More
Enzo Life Sciences Lovastatin (10mg). CAS: 75330-75-5
SDP
Supplier:  Enzo Life Sciences BMLG2260010
Encompass_Preferred
Inhibits cholesterol/isoprenoid biosynthesis by inhibition of HMG-CoA reductase (Ki for acid form is 1nM) and blocks protein isoprenylation and reduces plasma cholesterol levels in humans. Causes cells to arrest early in the G1 phase. Induces apoptosis in cancer cells. Alternative name: Mevinolin, 6-alpha-Methylcompactin. Solubility: Soluble in DMSO (>25mg/ml) or 100% ethanol (>35mg/ml); insoluble in water. Long Term Storage: -20°C.
Catalog No. 50-201-0858