Learn More
AdipoGen Lovastatin
SDP
Supplier:  AdipoGen AGCN20051M025
Encompass_Preferred
Chemical. CAS 75330-75-5. Formula C24H36O5. MW 404.6. Synthetic. Statin compound. Potent competitive HMG-CoA reductase inhibitor. Anti-hypercholesterolemic agent. Cholesterol/isoprenoid biosynthesis inhibitor. Blocks the production of mevalonate, a critical compound in the production of cholesterol and isoprenoids. Inhibits the isoprenylation of Ras and Rho family GTPases. Causes cell cycle arrest in G1 and G2/M phases through modulation of proteasome in cancer cell lines. Smooth muscle cell proliferation inhibitor. Apoptosis inducer. Anticancer compound. Anti-adhesive, immunomodulatory and anti-inflammatory compound. Stimulates bone formation. Increases cellular resistance to anticancer agents such as doxorubicin and etoposide Prod. No. AG-CR1-3572 http//www.adipogen.com/ag-cr1-3572/etoposide.html . Suppresses ICAM-1-LFA-1 interactions, which blocks virus replication and infection. Anti-hypertensive agent. Suppresses TNF-induced NF-kappaB activation.
Catalog No. 50-256-2681