Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Lovastatin | 75330-75-5 | 404.54 | 250 MG
SDP

Catalog No. 5000389568
Encompass_Preferred

Lovastatin is a cell-permeable HMG-CoA reductase inhibitor used to lower cholesterol. In its active form, it inhibits the enzyme HMG-CoA reductase, significantly reducing apolipoprotein-B-containing lipoproteins like LDL cholesterol and plasma triglycerides, while slightly increasing HDL cholesterol. It also effectively reduces cell viability and induces apoptosis in HepG2 cells.

  • Inhibits HMG-CoA reductase
  • Reduces HepG2 cell viability at 10 μM
  • Induces HepG2 cell apoptosis at 10 μM
  • Purity: 99.75%
  • Appearance: Solid
  • Color: Off-white to light yellow
  • Molecular weight: 404.54
  • Formula: C24H36O5
  • Targets: HMG-CoA reductase (HMGCR), autophagy, ferroptosis
  • Pathways: Metabolic enzyme/protease, autophagy, apoptosis

Catalog No. 50-003-89568 Supplier Medchemexpress LLC Supplier No. 250MGHYN0504
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Lovastatin is a cell-permeable HMG-CoA reductase inhibitor used to lower cholesterol. In its active form, it inhibits the enzyme HMG-CoA reductase, significantly reducing apolipoprotein-B-containing lipoproteins like LDL cholesterol and plasma triglycerides, while slightly increasing HDL cholesterol. It also effectively reduces cell viability and induces apoptosis in HepG2 cells.

  • Inhibits HMG-CoA reductase
  • Reduces HepG2 cell viability at 10 μM
  • Induces HepG2 cell apoptosis at 10 μM
  • Purity: 99.75%
  • Appearance: Solid
  • Color: Off-white to light yellow
  • Molecular weight: 404.54
  • Formula: C24H36O5
  • Targets: HMG-CoA reductase (HMGCR), autophagy, ferroptosis
  • Pathways: Metabolic enzyme/protease, autophagy, apoptosis

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.