Learn More
Medchemexpress LLC Lovastatin-d9 | 75330-75-5 | 413.60 | 5 MG
SDP
Catalog No. 5000389980
Encompass_Preferred
Lovastatin-d9 is a deuterium-labeled Lovastatin, functioning as a cell-permeable HMG-CoA reductase inhibitor used to lower cholesterol.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Contains stable heavy isotopes incorporated for drug development quantitation
  • Deuteration can impact pharmacokinetic and metabolic profiles
Catalog No. 50-003-89980 Supplier Medchemexpress LLC Supplier No. HYN0504S15MG
Lovastatin-d9 is a deuterium-labeled Lovastatin, functioning as a cell-permeable HMG-CoA reductase inhibitor used to lower cholesterol.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Contains stable heavy isotopes incorporated for drug development quantitation
  • Deuteration can impact pharmacokinetic and metabolic profiles