Learn More
Cayman Chemical Lovastatin Hydroxy Acid sodi
SDP
Supplier:  Cayman Chemical 1001033910 MG
Encompass_Preferred
A potent and competitive HMG-CoA reductase inhibitor (Ki = 0.6 nM); is the more potent, open ring, hydroxy acid form of its prodrug, lovastatin
Catalog No. 50-288-9150