Learn More
Cayman Chemical Lovastatin Hydroxy Acid sodi
SDP
Catalog No. 502889199
Encompass_Preferred
A potent and competitive HMG-CoA reductase inhibitor (Ki = 0.6 nM); is the more potent, open ring, hydroxy acid form of its prodrug, lovastatin
Catalog No. 50-288-9199 Supplier Cayman Chemical Supplier No. 1001033925 MG
A potent and competitive HMG-CoA reductase inhibitor (Ki = 0.6 nM); is the more potent, open ring, hydroxy acid form of its prodrug, lovastatin