Learn More
Cayman Chemical Lovastatin Hydroxy Acid (sodium salt), Purity: ≥95%, CAS: 75225-50-2, 1 mg
SDP
Catalog No. 502888481
Encompass_Preferred
A potent and competitive HMG-CoA reductase inhibitor (Ki = 0.6 nM); is the more potent, open ring, hydroxy acid form of its prodrug, lovastatin
Catalog No. 50-288-8481 Supplier Cayman Chemical Supplier No. 100103391 MG
A potent and competitive HMG-CoA reductase inhibitor (Ki = 0.6 nM); is the more potent, open ring, hydroxy acid form of its prodrug, lovastatin