Learn More
Medchemexpress LLC Lovastatin hydroxy acid sodium | 75225-50-2 | MFCD00870475 | 99.5% | 444.54 | C24H37NaO6 | 5MG
SDP
Catalog No. 5000237316
Encompass_Preferred
Lovastatin hydroxy acid sodium is the sodium salt of the active hydroxy acid form of lovastatin and a potent HMG-CoA reductase inhibitor (Ki = 0.6 nM). It is supplied as a high-purity analytical standard for biochemical research, enzymology, and inhibitor profiling.
  • High purity for analytical and research applications.
  • Potent HMG-CoA reductase inhibition (Ki = 0.6 nM).
  • Sodium salt form improves aqueous solubility compared with the lactone.
  • Available in small quantities suitable for assay development.
  • Includes supporting documentation such as datasheet, certificate of analysis, and safety data sheet.
Catalog No. 50-002-37316 Supplier Medchemexpress LLC Supplier No. HY1236725MG
Lovastatin hydroxy acid sodium is the sodium salt of the active hydroxy acid form of lovastatin and a potent HMG-CoA reductase inhibitor (Ki = 0.6 nM). It is supplied as a high-purity analytical standard for biochemical research, enzymology, and inhibitor profiling.
  • High purity for analytical and research applications.
  • Potent HMG-CoA reductase inhibition (Ki = 0.6 nM).
  • Sodium salt form improves aqueous solubility compared with the lactone.
  • Available in small quantities suitable for assay development.
  • Includes supporting documentation such as datasheet, certificate of analysis, and safety data sheet.