Learn More
Abcam Lovastatin (Mevinolin), HMG-CoA reductase inhibitor, 10MG
Catalog No. 502707424
Encompass_Preferred
MW 404.5 Da, Purity >98%. Potent, competitive HMG-CoA reductase inhibitor (Ki = 0.6 nM). Blood-brain barrier permeable. Inhibits in vivo and in vitro cholesterol biosynthesis. Also reduces α-synuclein levels in vitro and in vivo.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-7424 Supplier Abcam Supplier No. AB12061410MG
MW 404.5 Da, Purity >98%. Potent, competitive HMG-CoA reductase inhibitor (Ki = 0.6 nM). Blood-brain barrier permeable. Inhibits in vivo and in vitro cholesterol biosynthesis. Also reduces α-synuclein levels in vitro and in vivo.
The product is subject to the following: Abcam Restricted Use Statement