Learn More
Lovibond™ 10 mm Path Length Glass Cells
Catalog No. 09053450 Shop All Tintometer Group Products
Change view
Click to view available options
Type:
Glass Cells
Catalog No. Type
09-053-450 Glass Cells
1 options
Catalog No. 09-053-450 Supplier Lovibond™ Supplier No. 606810
Add to Cart
Add to Cart
A precision fused cuvette made from optical glass to the highest standards.
  • 10mm path length
  • For measuring the color of varnishes, drying oils, resins, nitro-cellulose, lacquers, fatty acids and similar transparent colored liquids
  • Complies with ASTM D-1544 and AOCS Method Td 1a
Specifications
Type Glass Cells
For Use With (Equipment) Comparator 2000+, comparator 3000 with non-heated Samples