Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lovibond™ Alka-M High Range Colorimtere Tablets Reagent

Catalog No. 09534604 Shop All Tintometer Group Products
Change view
Click to view available options
Range:
5 to 500ppm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Range
09-534-604 5 to 500ppm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-534-604 Supplier Lovibond™ Supplier No. 513240BT
Add to Cart
Add to Cart

Lovibond Alka-M High Range Photometer Tablets, Alka-M HR Tablet, Range: 5 to 500 ppm, Orange, Composition: Adipic Acid 10 to 20%, non-hazardous components 80 to 90%, Format: Tablet, For Use With: Water analysis, MD600 colorimeter, Spectrodirect & Multidirect spectrophotometer, XD7000, and AquaPRO6, Method: Acid Indicator

Lovibond Alka-M High Range Photometer Tablets, Alka-M HR Tablet, Range: 5 to 500ppm, Orange, Composition: Adipic Acid 10 to 20%, non-hazardous components 80 to 90%, Format: Tablet, For Use With: Water analysis, MC500 colorimeter, SP600 spectrophotometer, AquaPRO6, Method: Acid Indicator

Specifications

Color Orange
Composition Adipic Acid 10 to 20%, non-hazardous components 80 to 90%
pH Range 3.1
Format Tablet
For Use With (Application) Water analysis
For Use With (Equipment) MC500 colorimeter, SP600 spectrophotometer, AquaPRO6
Range 5 to 500ppm
Storage Requirements 20°C
Test Method Acid Indicator
Type Alka-M HR Tablet
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.