Learn More
Lovibond™ Aqua Comparitor Color Discs: Iron
Catalog No. 09053150 Shop All Tintometer Group Products
Change view
Click to view available options
Range:
0 to 1.8mg/L
0.05 to 1mg/L
1 to 10mg/L
Type:
Iron HR Color Disc
Iron TPTZ Color Disc
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Range Type
09-053-150 0.05 to 1mg/L Iron HR Color Disc
09-053-160 1 to 10mg/L Iron HR Color Disc
09-053-177 0 to 1.8mg/L Iron TPTZ Color Disc
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 09-053-150 Supplier Lovibond™ Supplier No. 146220
Only null left
Add to Cart
Add to Cart
For testing water with the Aqua Comparator
Economical replacement discs make it easy to test for iron using the Aqua Comparator body. Just slip the disc into the comparator and add the related reagent (available separately).
Specifications
For Use With (Application) Testing water with the Aqua comparator™
For Use With (Equipment) Aqua comparator™
Range 0.05 to 1mg/L
Type Iron HR Color Disc