Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lovibond™ Bromocresol Purple Tablets

Catalog No. 09534623 Shop All Tintometer Group Products
Change view
Click to view available options
pH Range:
5.2 to 6.8
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. pH Range
09-534-623 5.2 to 6.8
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-534-623 Supplier Lovibond™ Supplier No. 511730
Add to Cart
Add to Cart

Convenient blister packaging allows user to push tablet through foil directly into sample

  • Each tablet hermetically sealed in individual aluminum foil pocket that protects it from environmental conditions for longer shelf life
  • For use with pH Aqua Comparator™ Test Kit
  • For pH measurement in water samples
  • pH Range: 5.2 to 6.8

Specifications

Color Orange
Composition Potassium Chloride >95%, Bromothymol purple 0.1 to 1%, Sodium Thiosulphate <2.5%
pH Range 5.2 to 6.8
Format Tablet
For Use With (Application) Water analysis
For Use With (Equipment) Colorimeter and spectrophotometer
Storage Requirements 20°C
Test Method Bromocresol Purple
Type Bromocresol Purple Tablet
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.