Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lovibond™ CHECKIT™ Comparator Test Kits: Chlorine-Total

Catalog No. 09053107 Shop All Tintometer Group Products
Change view
Click to view available options
Range:
0 to 3.5mg/L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Range
09-053-107 0 to 3.5mg/L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-053-107 Supplier Lovibond™ Supplier No. 147051
Add to Cart
Add to Cart

Includes

  • Aqua Comparator Housing
  • Aqua Comparator Disc
  • DPD Total Reagent, 5mL sample (100 pack)
  • 2 x 13.5mm Sample Cells
  • Tablet Crusher
  • Instructions
  • Carrying Case

Accurate test kits for routine water analysis

  • Easy to use: add reagent to sample cell, rotate disc until color matches sample, and read concentration value

Specifications

Accuracy ±5%
For Use With (Application) Routine water analysis
Includes Aqua comparator™ housing, aqua comparator™ disc, DPD total reagent, 5mL sample, 2 x 13.5mm sample cells, tablet crusher, instructions, carrying case
No. of Tests 100
Range 0 to 3.5mg/L
Type Chlorine Free and Total Test Kit
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.