Learn More
Lovibond™ CHECKIT™ Comparator Test Kits: Copper-Free/Total
Catalog No. 09053112 Shop All Tintometer Group Products
Change view
Click to view available options
Range:
0 to 1mg/L
Catalog No. Range
09-053-112 0 to 1mg/L
1 options
Catalog No. 09-053-112 Supplier Lovibond™ Supplier No. 147440
Add to Cart
Add to Cart
Includes
  • Aqua Comparator Housing
  • Aqua Comparator Disc
  • Copper No. 1 Tablets (100 pack)
  • Copper No. 2 Tablets (100 pack)
  • 2 x 13.5mm Sample Cells
  • Tablet Crusher
  • Instructions
  • Carrying Case
Accurate test kits for routine water analysis
  • Easy to use: add reagent to sample cell, rotate disc until color matches sample, and read concentration value
Specifications
Accuracy ±5%
For Use With (Application) Routine water analysis
Includes Aqua comparator™ housing, aqua comparator™ disc, copper No. 1 tablets, copper No. 2 tablets, 2 x 13.5mm sample cells, tablet crusher, instructions, carrying case
No. of Tests 100
Range 0 to 1mg/L
Type Copper Free Total Test Kit