Learn More
Description
Includes
- Aqua Comparator Housing
- Aqua Comparator Disc
- Iron LR Tablets (100 pack)
- Iron (II) LR Tablets (100 pack)
- 2 x 13.5mm Sample Cells
- Tablet Crusher
- Instructions
- Carrying Case
- Easy to use: add reagent to sample cell, rotate disc until color matches sample, and read concentration value
Specifications
Specifications
| Accuracy | ±5% |
| For Use With (Application) | Routine water analysis |
| Includes | Aqua comparator™ housing, aqua comparator™ disc, iron LR tablets, iron (II) LR tablets, 2 x 13.5mm sample cells, tablet crusher, instructions, carrying case |
| No. of Tests | 100 |
| Range | 0 to 1mg/L |
| Type | LR Iron Test Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.