Learn More
Description
Lovibond Chloride T2 Tablets, Range: 0.5 to 25mg/L, White, Composition: Potassium Monopersulfate Triple salt 5 to 10%, Silver Nitrate <2.5%, Boric Acid >50%, non-hazardous components <20%, Format: Tablet, For Use With: Water analysis, MC500 colorimeter and SP600 spectrophotometer
- These tablet reagents provide a safe and reliable method for chloride analysis in wastewater treatment applications.
- The tablet form ensures high accuracy and enables precise dosing, while the aluminium blister packaging provides durability.
- Chemical Method: Silver Nitrate / Turbidity
- Parameter: Chloride
- Measuring Range: 0.5 to 25 mg/L Cl-
- Packaging: Aluminium blister pack
- Quantity: 100 pc.
- Wavelength: 530 nm
- Cuvette Requirement: ø 24 mm
- Standard: DIN 38405
Specifications
Specifications
| Color | White |
| Composition | Potassium Monopersulfate Triple salt 5 to 10%, Silver Nitrate <2.5%, Boric Acid >50%, non-hazardous components <20% |
| pH Range | 4 |
| Format | Tablet |
| For Use With (Application) | Water analysis |
| For Use With (Equipment) | MC500 colorimeter and SP600 spectrophotometer |
| Range | 0.5 to 25mg/L |
| Storage Requirements | 20°C |
| Test Method | Silver Nitrate/Turbidity |
| Type | Chloride T2 Tablet |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.