Learn More
Lovibond™ COD Tube Tests, HR, Mercury Free
Catalog No. 09053313 Shop All Tintometer Group Products
Change view
Click to view available options
Range:
0 to 15000mg/L
Catalog No. Range
09-053-313 0 to 15000mg/L
1 options
Catalog No. 09-053-313 Supplier Lovibond™ Supplier No. 2420712
Add to Cart
Add to Cart
Premeasured unit dose reagent for water quality testing
  • For use in SC400™ Colorimeters and Aqua Comparator™ Test Kits
  • Also suitable for use in Hach™ instruments and test kits
  • Easy-to-open, sealed vials eliminate risk of cross-contamination
  • Extended shelf life of up to five years
For Municipal Wastewater Testing; Industrial Wastewater and Process Control Testing; Environmental Water Testing
Compliance
US EPA-accepted test method
Specifications
Certifications/Compliance US EPA
For Use With (Application) Municipal wastewater testing, industrial wastewater and process control testing, environmental water testing
For Use With (Equipment) SC400 colorimeters and Aqua comparator™ test kits, Hach™ instruments
No. of Tests 25
Range 0 to 15000mg/L
Test Method Dichromate/H2SO4