Learn More
Lovibond™ COD Tube Tests; MR; Mercury Free
Catalog No. p-4780382 Shop All Tintometer Group Products
Change view
Click to view available options
No. of Tests:
25
Catalog No. No. of Tests
09-053-311 25
1 options
Catalog No. 09-053-311 Supplier Lovibond™ Supplier No. 2420711
Only null left
Add to Cart
Add to Cart
Premeasured unit dose reagent for water quality testing
  • For use in SC400™ Colorimeters and Aqua Comparator™ Test Kits
  • Also suitable for use in Hach™ instruments and test kits
  • Easy-to-open, sealed vials eliminate risk of cross-contamination
  • Extended shelf life of up to five years
For Municipal Wastewater Testing; Industrial Wastewater and Process Control Testing; Environmental Water Testing
Compliance
US EPA-accepted test method
Specifications
Certifications/Compliance US EPA
For Use With (Application) Municipal wastewater testing, industrial wastewater and process control testing, environmental water testing
For Use With (Equipment) SC400 colorimeters and Aqua comparator™ test kits, Hach™ instruments
No. of Tests 25
Range 0 to 1500mg/L
Test Method Dichromate/H2SO4