Specifications
| Certifications/Compliance | ASTM D-1544, AOCS Td 1a |
| Type | Color of Varnish l Disc |
| For Use With (Equipment) | Varnish comparator kits 705-V2, 705-VA2 or 705-VA4 |
| Range | 1 to 9 |