Description
Use with MC500, MD600, SC450, MD100 colorimeters, SP600 spectrophotometer, Pool Control 9, AquaPRO3 Pool and AquaPRO6 Pool photometers and Scuba IIElectronic Pool Tester
Specifications
Specifications
| Type | DPD No. 1 Colorimeter Tablet Reagent |
| For Use With (Equipment) | MC500, MD600, SC450, MD100 colorimeters, SP600 spectrophotometer, Pool Control 9, AquaPRO3 Pool and AquaPRO6 Pool photometers and Scuba IIElectronic Pool Tester |
| Includes | 100 Tablets/Pack |