Learn More
Lovibond™ Comparator Disc for 705 Comparator
Catalog No. 15398101E Shop All Tintometer Group Products
Change view
Click to view available options
Type:
4/78 Comparator Disc
Catalog No. Type
15-398-101E 4/78 Comparator Disc
1 options
Catalog No. 15-398-101E Supplier Lovibond™ Supplier No. 247800
Specifications
Type 4/78 Comparator Disc
For Use With (Application) Measuring dyed aviation gasoline per Dyed Aviation Gasoline ASTM D 2392 (Blue, Green, Brown, Purple Min/Max)
For Use With (Equipment) 705 Comparator