Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lovibond™ DPD No. 1 HR Tablets

Catalog No. 09534606 Shop All Tintometer Group Products
Change view
Click to view available options
Range:
0.01 to 10ppm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Range
09-534-606 0.01 to 10ppm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-534-606 Supplier Lovibond™ Supplier No. 511500BT
Add to Cart
Add to Cart

Determine the amount of free chlorine in your hot tub, swimming pool, or spa water with these foil-packaged, fast-acting tablets. Lovibond™ DPD No. 1 HR Tablets are foil-packaged tablets for consistent and reliable reagent delivery.

  • Simple, time-saving and practical
  • Foil packaged test tablets remain the most consistent and reliable reagent delivery system available
  • Each tablet is hermetically sealed within an individually formed foil cup

Specifications

Color White
Composition N,N-diethylbenzene-1,4-diammonium Sulphate 2.5 to 5%, Citric Acid 0.1 to 1%, non-hazardous components 20 to 35%, Sodium cyclamate 60 to 70%, Polyvinyl Alcohol ≤0.1%, Di-Sodium Dihydrogenethylenediaminetetraacetate ≤2.5%
pH Range 6.3
Format Tablet
For Use With (Application) Free chlorine analysis in water
For Use With (Equipment) SC450 chlorine 2 and 3 colorimeters, MC500 colorimeter, SP600 spectrophotometer, AquaPRO3 and PRO6 Pool photometers
Range 0.01 to 10ppm
Storage Requirements 20°C
Test Method DPD
Type DPD No. 1 HR Tablet
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.