Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Lovibond™ DPD No. 3 Rapid Tablets

Catalog No. 09534616 Shop All Tintometer Group Products
Change view
Click to view available options
Range:
0 to 1mg/L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Range
09-534-616 0 to 1mg/L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 09-534-616 Supplier Lovibond™ Supplier No. 511290BT
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Convenient blister packaging allows user to push tablet through foil directly into sample

DPD Tablet No. 1 is used to measure free chlorine; after that, DPD Tablet No. 3 is used to measure total chlorine.

  • Each tablet hermetically sealed in individual aluminum foil pocket that protects it from environmental conditions for longer shelf life
  • For use with Chlorine Aqua Comparator™ Test Kits
  • Range: 0 to 1mg/L

Specifications

Color White
Composition Potassium Iodide 10 to 20%, non-hazardous components 80 to 90%, Cellulose 0.1 to 1%
pH Range 6.3
Format Tablet
For Use With (Application) Water analysis
For Use With (Equipment) Colorimeter and spectrophotometer
Range 0 to 1mg/L
Test Method DPD
Type DPD No. 3 Rapid Tablet
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.