Learn More
Lovibond™ Gardner Color Scale Based on Comparator System 2000
Catalog No. 09053481 Shop All Tintometer Group Products
Change view
Click to view available options
Type:
Gardner Color Scale
Catalog No. Type
09-053-481 Gardner Color Scale
1 options
Catalog No. 09-053-481 Supplier Lovibond™ Supplier No. 433340
Only null left
Add to Cart
Add to Cart
Request a Quote
A flexible, modular system for visual color grading.
  • Type: AF 334
  • Range: 1-18 units
  • Includes: Lovibond Comparator 2000+ with Daylight 2000 Lighting Unit, Gardner discs 4/30 AS and 4/30 BS, and (2) W680/OG/10mm path length fused glass cells
Compliance
Conforms to ASTM D1544, ISO4630 Standards
Specifications
Certifications/Compliance Conforms to ASTM D1544, ISO4630 Standards
Description Gardner Color Scale Based on Comparator System 2000
Type Gardner Color Scale
For Use With (Application) A flexible, modular system for visual color grading.
Includes Lovibond Comparator 2000+ with Daylight 2000 Lighting Unit, Gardner discs 4/30 AS and 4/30 BS, and (2) W680/OG/10mm path length fused glass cells
Model AF 334
Range 1 to 18 units